Assay Details
| Gene Symbol: LOC4343510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789698.2 | XP_015645184.1 | 3 - 4 | 951 | 98 | |
| XM_015789699.2 | XP_015645185.1 | 3 - 4 | 904 | 98 | ||
More Information
Set Membership: |
|

LOC4343510
Genomic Map