Assay Details
| Gene Symbol: LOC4343576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792027.2 | XP_015647513.1 | 1 - 2 | 1385 | 65 | |
| GenBank mRNA | AK067121.1 | - | 1 - 2 | 84 | 65 | |
| AK101830.1 | - | 1 - 2 | 1361 | 65 | ||
More Information
Set Membership: |
|

LOC4343576
Genomic Map