Assay Details
| Gene Symbol: LOC4343576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792027.2 | XP_015647513.1 | 4 - 5 | 1748 | 77 | |
| GenBank mRNA | AK067121.1 | - | 4 - 5 | 447 | 77 | |
| AK101830.1 | - | 4 - 5 | 1724 | 77 | ||
More Information
Set Membership: |
|

LOC4343576
Genomic Map