Assay Details
| Gene Symbol: LOC4343790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789728.1 | XP_015645214.1 | 1 - 2 | 452 | 77 | |
| GenBank mRNA | AK066349.1 | - | 1 - 2 | 298 | 77 | |
More Information
Set Membership: |
|

LOC4343790
Genomic Map