Assay Details
| Gene Symbol: LOC9267772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK065600.1 | - | 4 - 5 | 847 | 145 | |
| AK103165.1 | - | 4 - 5 | 847 | 145 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC9267772| Gene Symbol: LOC9267772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK065600.1 | - | 4 - 5 | 847 | 145 | |
| AK103165.1 | - | 4 - 5 | 847 | 145 | ||
Set Membership: |
|