Assay Details
Gene Symbol: LOC9267772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789467.2 | XP_015644953.1 | 8 - 9 | 1554 | 71 | |
GenBank mRNA | AK065600.1 | - | 8 - 9 | 1336 | 71 | |
AK103165.1 | - | 8 - 9 | 1336 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
