Assay Details
| Gene Symbol: LOC4343817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790670.2 | XP_015646156.1 | 1 | 546 | 83 | |
| GenBank mRNA | AK062681.1 | - | 1 | 552 | 83 | |
| AK105814.1 | - | 1 | 552 | 83 | ||
More Information
Set Membership: |
|

LOC4343817
Genomic Map