Assay Details
| Gene Symbol: LOC4343963 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790857.2 | XP_015646343.1 | 1 | 646 | 74 | |
| GenBank mRNA | AK064628.1 | - | 1 | 652 | 74 | |
More Information
Set Membership: |
|

LOC4343963
Genomic Map