Assay Details
| Gene Symbol: LOC4344194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791268.2 | XP_015646754.1 | 1 - 2 | 576 | 121 | |
| XM_015791269.2 | XP_015646755.1 | 1 - 2 | 576 | 121 | ||
| XM_015791270.2 | XP_015646756.1 | 1 - 2 | 577 | 121 | ||
| GenBank mRNA | AK100642.1 | - | 1 - 2 | 404 | 121 | |
More Information
Set Membership: |
|

LOC4344194
Genomic Map