Assay Details
| Gene Symbol: LOC4344194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791268.2 | XP_015646754.1 | 4 - 5 | 838 | 150 | |
| XM_015791270.2 | XP_015646756.1 | 4 - 5 | 839 | 150 | ||
| GenBank mRNA | AK070340.1 | - | 3 - 4 | 351 | 150 | |
| AK100642.1 | - | 4 - 5 | 666 | 150 | ||
More Information
Set Membership: |
|

LOC4344194
Genomic Map