Assay Details
| Gene Symbol: LOC4344194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791270.2 | XP_015646756.1 | 6 - 7 | 1115 | 75 | |
| GenBank mRNA | AK100642.1 | - | 6 - 7 | 942 | 75 | |
More Information
Set Membership: |
|

LOC4344194
Genomic Map