Assay Details
| Gene Symbol: LOC4344194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791268.2 | XP_015646754.1 | 8 - 9 | 1660 | 106 | |
| XM_015791269.2 | XP_015646755.1 | 8 - 9 | 1654 | 106 | ||
| XM_015791270.2 | XP_015646756.1 | 7 - 8 | 1445 | 106 | ||
| GenBank mRNA | AK070340.1 | - | 7 - 8 | 1173 | 106 | |
| AK100642.1 | - | 7 - 8 | 1272 | 106 | ||
More Information
Set Membership: |
|

LOC4344194
Genomic Map