Assay Details
| Gene Symbol: LOC4344194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791268.2 | XP_015646754.1 | 9 - 10 | 1806 | 129 | |
| XM_015791269.2 | XP_015646755.1 | 9 - 10 | 1800 | 129 | ||
| XM_015791270.2 | XP_015646756.1 | 8 - 9 | 1591 | 129 | ||
| GenBank mRNA | AK070340.1 | - | 8 - 9 | 1319 | 129 | |
| AK100642.1 | - | 8 - 9 | 1418 | 129 | ||
More Information
Set Membership: |
|

LOC4344194
Genomic Map