Assay Details
| Gene Symbol: LOC4344256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791676.2 | XP_015647162.1 | 8 - 9 | 698 | 140 | |
| XM_015791677.2 | XP_015647163.1 | 8 - 9 | 652 | 140 | ||
| GenBank mRNA | AK072839.1 | - | 7 - 8 | 697 | 140 | |
| AK106176.1 | - | 8 - 9 | 629 | 140 | ||
More Information
Set Membership: |
|

LOC4344256
Genomic Map