Assay Details
| Gene Symbol: LOC4344256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791676.2 | XP_015647162.1 | 9 - 10 | 757 | 150 | |
| XM_015791677.2 | XP_015647163.1 | 9 - 10 | 711 | 150 | ||
| GenBank mRNA | AK072839.1 | - | 8 - 9 | 756 | 150 | |
| AK106176.1 | - | 9 - 10 | 688 | 150 | ||
More Information
Set Membership: |
|

LOC4344256
Genomic Map