Assay Details
| Gene Symbol: LOC4344356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789763.2 | XP_015645249.1 | 6 - 7 | 844 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4344356| Gene Symbol: LOC4344356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789763.2 | XP_015645249.1 | 6 - 7 | 844 | 64 | |
Set Membership: |
|