Assay Details
| Gene Symbol: LOC4344819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794437.2 | XP_015649923.1 | 19 - 20 | 6763 | 96 | |
| GenBank mRNA | AK058948.1 | - | 3 - 4 | 377 | 96 | |
| AK121489.1 | - | 10 - 11 | 1187 | 96 | ||
More Information
Set Membership: |
|

LOC4344819
Genomic Map