Assay Details
| Gene Symbol: LOC4345334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795377.2 | XP_015650863.1 | 7 - 8 | 1170 | 116 | |
| GenBank mRNA | AK059591.1 | - | 2 - 3 | 169 | 116 | |
| AK066618.1 | - | 7 - 8 | 1158 | 116 | ||
| BT129367.1 | - | 2 - 3 | 191 | 116 | ||
More Information
Set Membership: |
|

LOC4345334
Genomic Map