Assay Details
| Gene Symbol: LOC4345404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795075.2 | XP_015650561.1 | 3 - 4 | 797 | 152 | |
| XM_015795076.2 | XP_015650562.1 | 3 - 4 | 770 | 152 | ||
| XM_015795077.2 | XP_015650563.1 | 3 - 4 | 760 | 152 | ||
| XM_015795078.2 | XP_015650564.1 | 3 - 4 | 797 | 152 | ||
More Information
Set Membership: |
|

LOC4345404
Genomic Map