Assay Details
Gene Symbol: LOC4345991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793050.2 | XP_015648536.1 | 1 - 2 | 300 | 114 | |
GenBank mRNA | AK099585.1 | - | 369 | 114 | ||
AK105183.1 | - | 1 - 2 | 276 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
