Assay Details
Gene Symbol: LOC4345991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015793050.2 | XP_015648536.1 | 2 - 3 | 400 | 75 | |
GenBank mRNA | AK105183.1 | - | 2 - 3 | 376 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
