Assay Details
| Gene Symbol: LOC4345993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795231.2 | XP_015650717.1 | 2 - 3 | 955 | 60 | |
| GenBank mRNA | AF095895.1 | - | 2 - 3 | 361 | 60 | |
| AK241395.1 | - | 2 - 3 | 815 | 60 | ||
| D14000.1 | - | 2 - 3 | 791 | 60 | ||
| FJ607153.1 | - | 2 - 3 | 687 | 60 | ||
More Information
Set Membership: |
|

LOC4345993
Genomic Map