Assay Details
| Gene Symbol: LOC4346134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793276.1 | XP_015648762.1 | 1 - 2 | 254 | 78 | |
| GenBank mRNA | AK100263.1 | - | 1 - 2 | 251 | 78 | |
| AY332477.1 | - | 1 - 2 | 233 | 78 | ||
| AY551921.1 | - | 2 - 3 | 418 | 78 | ||
| U78891.2 | - | 1 - 2 | 283 | 78 | ||
| Y15008.1 | - | 1 - 2 | 187 | 78 | ||
More Information
Set Membership: |
|

LOC4346134
Genomic Map