Assay Details
| Gene Symbol: LOC4346134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793276.1 | XP_015648762.1 | 6 - 7 | 587 | 85 | |
| GenBank mRNA | AK100263.1 | - | 6 - 7 | 584 | 85 | |
| AY551921.1 | - | 8 - 9 | 856 | 85 | ||
| U78891.2 | - | 6 - 7 | 616 | 85 | ||
| Y15008.1 | - | 6 - 7 | 520 | 85 | ||
More Information
Set Membership: |
|

LOC4346134
Genomic Map