Assay Details
| Gene Symbol: LOC4346201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792795.2 | XP_015648281.1 | 4 - 5 | 676 | 86 | |
| GenBank mRNA | AK064378.1 | - | 4 - 5 | 672 | 86 | |
| AK074001.1 | - | 4 - 5 | 684 | 86 | ||
More Information
Set Membership: |
|

LOC4346201
Genomic Map