Assay Details
| Gene Symbol: LOC4346210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794439.2 | XP_015649925.1 | 16 - 17 | 2931 | 69 | |
| GenBank mRNA | AK107688.1 | - | 1 - 2 | 279 | 69 | |
More Information
Set Membership: |
|

LOC4346210
Genomic Map