Assay Details
| Gene Symbol: LOC4346210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794439.2 | XP_015649925.1 | 17 - 18 | 3019 | 105 | |
| GenBank mRNA | AK107688.1 | - | 2 - 3 | 367 | 105 | |
More Information
Set Membership: |
|

LOC4346210
Genomic Map