Assay Details
| Gene Symbol: LOC9267891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756781.2 | XP_015612267.1 | 16 - 17 | 2080 | 149 | |
| XM_015756782.2 | XP_015612268.1 | 14 - 15 | 2436 | 149 | ||
| XM_015756784.2 | XP_015612270.1 | 14 - 15 | 1744 | 149 | ||
More Information
Set Membership: |
|

LOC9267891
Genomic Map