Assay Details
| Gene Symbol: LOC9267891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756781.2 | XP_015612267.1 | 2 - 3 | 309 | 77 | |
| GenBank mRNA | AK106637.1 | - | 2 - 3 | 274 | 77 | |
More Information
Set Membership: |
|

LOC9267891
Genomic Map