Assay Details
| Gene Symbol: LOC9267891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756781.2 | XP_015612267.1 | 5 - 6 | 578 | 131 | |
| XM_015756782.2 | XP_015612268.1 | 3 - 4 | 934 | 131 | ||
| XM_015756784.2 | XP_015612270.1 | 3 - 4 | 242 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015756784.2 transcripts.
Set Membership: |
|

LOC9267891
Genomic Map