Assay Details
| Gene Symbol: LOC4347424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755423.2 | XP_015610909.1 | 1 | 1717 | 128 | |
| GenBank mRNA | AK105294.1 | - | 1 | 1651 | 128 | |
More Information
Set Membership: |
|

LOC4347424
Genomic Map