Assay Details
| Gene Symbol: LOC4347615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756454.1 | XP_015611940.1 | 3 - 4 | 699 | 88 | |
| GenBank mRNA | AK071023.1 | - | 3 - 4 | 700 | 88 | |
| AK105872.1 | - | 3 - 4 | 771 | 88 | ||
More Information
Set Membership: |
|

LOC4347615
Genomic Map