Assay Details
| Gene Symbol: LOC4347718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015755203.2 | XP_015610689.1 | 5 - 6 | 817 | 77 | |
| GenBank mRNA | AK119414.1 | - | 5 - 6 | 816 | 77 | |
More Information
Set Membership: |
|

LOC4347718
Genomic Map