Assay Details
| Gene Symbol: LOC4347728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757052.2 | XP_015612538.1 | 2 - 3 | 1295 | 91 | |
| GenBank mRNA | AK287606.1 | - | 2 - 3 | 1246 | 91 | |
More Information
Set Membership: |
|

LOC4347728
Genomic Map