Assay Details
| Gene Symbol: LOC4348242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757312.2 | XP_015612798.1 | 4 - 5 | 598 | 84 | |
| XM_015757313.2 | XP_015612799.1 | 5 - 6 | 694 | 84 | ||
| GenBank mRNA | AK068221.1 | - | 4 - 5 | 574 | 84 | |
More Information
Set Membership: |
|

LOC4348242
Genomic Map