Assay Details
| Gene Symbol: LOC4348629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758716.2 | XP_015614202.1 | 8 - 9 | 1176 | 122 | |
| GenBank mRNA | AK111779.1 | - | 8 - 9 | 1181 | 122 | |
More Information
Set Membership: |
|

LOC4348629
Genomic Map