Assay Details
| Gene Symbol: LOC4349071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759559.2 | XP_015615045.1 | 11 - 12 | 1595 | 81 | |
| GenBank mRNA | AK066499.1 | - | 11 - 12 | 1566 | 81 | |
More Information
Set Membership: |
|

LOC4349071
Genomic Map