Assay Details
| Gene Symbol: LOC4349071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759559.2 | XP_015615045.1 | 1 - 2 | 274 | 66 | |
| GenBank mRNA | AK061575.1 | - | 1 - 2 | 261 | 66 | |
| AK066499.1 | - | 1 - 2 | 245 | 66 | ||
More Information
Set Membership: |
|

LOC4349071
Genomic Map