Assay Details
| Gene Symbol: LOC4349488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757975.2 | XP_015613461.1 | 1 - 2 | 225 | 107 | |
| GenBank mRNA | AK071939.1 | - | 1 - 2 | 226 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015757975.2 transcripts.
Set Membership: |
|

LOC4349488
Genomic Map