Assay Details
| Gene Symbol: LOC4349488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757975.2 | XP_015613461.1 | 3 - 4 | 762 | 52 | |
| GenBank mRNA | AB101646.1 | - | 2 - 3 | 592 | 52 | |
| AK071939.1 | - | 3 - 4 | 763 | 52 | ||
More Information
Set Membership: |
|

LOC4349488
Genomic Map