Assay Details
| Gene Symbol: LOC4349488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757975.2 | XP_015613461.1 | 6 - 7 | 1601 | 71 | |
| GenBank mRNA | AB101646.1 | - | 5 - 6 | 1431 | 71 | |
| AK071939.1 | - | 6 - 7 | 1602 | 71 | ||
More Information
Set Membership: |
|

LOC4349488
Genomic Map