Assay Details
| Gene Symbol: LOC4349488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757975.2 | XP_015613461.1 | 9 - 10 | 2091 | 79 | |
| GenBank mRNA | AB101646.1 | - | 8 - 9 | 1921 | 79 | |
| AK071939.1 | - | 9 - 10 | 2092 | 79 | ||
More Information
Set Membership: |
|

LOC4349488
Genomic Map