Assay Details
| Gene Symbol: LOC4349884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762088.2 | XP_015617574.1 | 3 - 4 | 599 | 104 | |
| GenBank mRNA | AK072561.1 | - | 3 - 4 | 556 | 104 | |
More Information
Set Membership: |
|

LOC4349884
Genomic Map