Assay Details
| Gene Symbol: LOC4349954 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762022.2 | XP_015617508.1 | 7 - 8 | 1869 | 68 | |
| GenBank mRNA | AK061734.1 | - | 4 - 5 | 381 | 68 | |
| AK066941.1 | - | 7 - 8 | 1861 | 68 | ||
More Information
Set Membership: |
|

LOC4349954
Genomic Map