Assay Details
| Gene Symbol: LOC4351232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760014.2 | XP_015615500.1 | 2 - 3 | 546 | 101 | |
| GenBank mRNA | AB110185.1 | - | 2 - 3 | 202 | 101 | |
| AK062013.1 | - | 2 - 3 | 220 | 101 | ||
| AK067429.1 | - | 2 - 3 | 342 | 101 | ||
| AK109413.2 | - | 2 - 3 | 338 | 101 | ||
More Information
Set Membership: |
|

LOC4351232
Genomic Map