Assay Details
| Gene Symbol: LOC4351232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015760014.2 | XP_015615500.1 | 5 - 6 | 916 | 90 | |
| GenBank mRNA | AB110185.1 | - | 5 - 6 | 572 | 90 | |
| AK062013.1 | - | 5 - 6 | 590 | 90 | ||
| AK067429.1 | - | 5 - 6 | 712 | 90 | ||
| AK109413.2 | - | 5 - 6 | 708 | 90 | ||
More Information
Set Membership: |
|

LOC4351232
Genomic Map