Assay Details
| Gene Symbol: LOC4330077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767296.2 | XP_015622782.1 | 1 - 2 | 308 | 162 | |
| GenBank mRNA | AK069985.1 | - | 1 - 2 | 267 | 162 | |
| AK104033.1 | - | 1 - 2 | 235 | 162 | ||
| AK121244.1 | - | 1 - 2 | 250 | 162 | ||
More Information
Set Membership: |
|

LOC4330077
Genomic Map