Assay Details
| Gene Symbol: LOC4330077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767296.2 | XP_015622782.1 | 4 - 5 | 1085 | 94 | |
| GenBank mRNA | AK121244.1 | - | 4 - 5 | 1027 | 94 | |
More Information
Set Membership: |
|

LOC4330077
Genomic Map