Assay Details
Gene Symbol: LOC4330128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015768083.2 | XP_015623569.1 | 3 - 4 | 578 | 66 | |
XM_015768084.2 | XP_015623570.1 | 1 - 2 | 33 | 66 | ||
GenBank mRNA | AK108580.1 | - | 1 - 2 | 33 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015768084.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
