Assay Details
| Gene Symbol: LOC4330128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768082.2 | XP_015623568.1 | 5 - 6 | 1371 | 129 | |
| XM_015768083.2 | XP_015623569.1 | 5 - 6 | 1373 | 129 | ||
| XM_015768084.2 | XP_015623570.1 | 3 - 4 | 832 | 129 | ||
| GenBank mRNA | AK108580.1 | - | 3 - 4 | 832 | 129 | |
More Information
Set Membership: |
|

LOC4330128
Genomic Map